Learn More
Medchemexpress LLC Glycerol-d5 | 62502-71-0 | 99.1% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1659S6100MG
Encompass_Preferred
Glycerol-d5 is a deuterium labeled Glycerol used in sample preparation and gel formation for polyacrylamide gel electrophoresis.
  • For research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-13236