Learn More
Medchemexpress LLC Glycerol-d5 (Trihydroxypropane-d5) | 62502-71-0 | 99.1% | 50 MG
Supplier: Medchemexpress LLC HYB1659S650MG
Glycerol-d5 is a deuterium labeled glycerol used in sample preparation and gel formation for polyacrylamide gel electrophoresis. It is intended for research use only and is not sold to patients.
- Can be used as a tracer.
- Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.