Learn More
Medchemexpress LLC Glycerol phenylbutyrate | 611168-24-2 | 99.8% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB208750MG
Encompass_Preferred
Glycerol phenylbutyrate is a sigma-2 (σ2) receptor ligand with a pKi of 8.02. It is categorized as a new generation ammonia scavenger agent and is intended for research use only. Glycerol phenylbutyrate (GPB) shows potential for treating hyperammonemia and may offer therapeutic benefits for conditions such as chronic hepatic encephalopathy or other inherited metabolic disorders.
  • Sigma-2 (σ2) receptor ligand with a pKi of 8.02
  • New generation ammonia scavenger agent
  • Potential for treating hyperammonemia
  • May benefit chronic hepatic encephalopathy and other inherited metabolic disorders
Catalog No. 50-004-10337