Learn More
Medchemexpress LLC Glycerol (standard) | 56-81-5 | 98.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1659R100MG
Encompass_Preferred
Glycerol (Standard) is the analytical standard of Glycerol, intended for research and analytical applications. It is used in sample preparation and gel formation for polyacrylamide gel electrophoresis. This product, an analytical standard, serves as a reference standard with a supplied assay and is commonly employed in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Used as an analytical standard.
  • Suitable for research and analytical applications.
  • Employed in sample preparation and gel formation for polyacrylamide gel electrophoresis.
  • Commonly used in qualitative, quantitative, and methodological research experiments including HPLC, GC, and MS.
Catalog No. 50-004-13233