Learn More
Cayman Chemical GlycIn-13C 250mg
SDP
Supplier:  Cayman Chemical 35130250 MG
Encompass_Preferred
An internal standard for the quantification of glycine by GC- or LC-MS
Catalog No. 50-291-0469