Learn More
Medchemexpress LLC Glycine-13C2 | 67836-01-5 | 99.9% | 77.05 | 13C2H5NO2 | 25mg
SDP
Supplier:  Medchemexpress LLC HYY0966S325MG
Encompass_Preferred
Glycine-13C2 is the 13C-labeled Glycine Glycine is an inhibitory neurotransmitter in the CNS and also acts as a co-agonist along with glutamate facilitating an excitatory potential at the glutaminergic N-methyl-D-aspartic acid (NMDA) receptors
Catalog No. 50-002-67775