Learn More
AG Scientific Inc Glycine, 5 KG
Supplier:  AG Scientific Inc G25855KG
Encompass_Preferred
Glycine is the simplest amino acid and is amphoteric. It can either be protonating or protonated, depending on pH. Because of this property, it is used for buffers.Glycine is an inhibitory neurotransmitter in CNS and modulates excitatory neurotransmission by potentiating the action of glutamate at N-methyl-D-aspartate (NMDA) receptors. In addition, it has been shown that glycine modulates macrophage effector functions implicated in CNS inflammation and in other related inflammatory conditions.CAS Number: 56-40-6Molecular Weight: 75.07 DaChemical Formula: C2H5NO2Solubility: Soluble in water (100 mg/ml), and acetone (slightly soluble). Insoluble in alcohol, and ether.Storage Temperature: Room TemperatureResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-7041