Learn More
Medchemexpress LLC Glycine, 5-oxo-L-prolyl- | 29227-88-1 | 98.6% | 186.17 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP4859
Encompass_Preferred
Pyr-Gly is an inhibitory peptide that inhibits the proliferation of retinal pigment epithelium (RPE) cells and fibroblasts. It is supplied as a white to off-white solid.
  • Inhibitory peptide
  • Inhibits proliferation of retinal pigment epithelium (RPE) cells and fibroblasts
  • 98.6% purity (HPLC)
  • Molecular weight of 186.17
  • For research use only
Catalog No. 50-004-00281