Learn More
Medchemexpress LLC Glycine (aminoacetic acid) | 56-40-6 | 100.0% | 75.07 g/mol | C2H5NO2 | 50mg
SDP
Supplier:  Medchemexpress LLC HYY0966R50MG
Encompass_Preferred
Glycine (Standard) is the analytical standard of Glycine This product is intended for research and analytical applications Glycine is an inhibitory neurotransmitter in the CNS and also acts as a co-agonist along with glutamate facilitating an excitatory potential at the glutaminergic N-methyl-D-aspartic acid (NMDA) receptors Glycine is orally active Glycine can be used to study cell protection cancer neurological diseases and angiogenesis[1][2][3][4][5][6]
Catalog No. 50-002-68095