Learn More
Alkali Scientific Glycine, Free Base, 500 G
SDP
Supplier:  Alkali Scientific GLY005CP
Encompass
Glycine, Free Base, 500g, is a standard-grade amino acid used in a variety of laboratory and biochemical applications. Glycine is crucial for protein synthesis, enzyme function, and neurotransmission. It is often used in cell culture media to support cell growth, as well as in buffer preparations for biochemical assays. This 500g package provides a convenient size for smaller-scale laboratory research and experimental applications. Glycine is also used in peptide synthesis, metabolic studies, and the preparation of reagents and buffers for use in clinical and pharmaceutical research. This product ensures high purity and consistent performance in laboratory experiments.
Catalog No. 50-006-59688