Learn More
Medchemexpress LLC Glycine, L-arginylglycyl-L-tyrosyl-L-seryl-L-leucyl- | 59587-18-7 | 99.4% | 651.71 | 5 MG

Supplier: Medchemexpress LLC HYP39355MG
Arg-Gly-Tyr-Ser-Leu-Gly corresponds to the sequence of residues from 21 through 26 in lysozyme. It functions as a substrate for protein kinase and is phosphorylated at the serine residue by protein kinase, making it useful in biochemical research.
- Appears as a white to off-white solid
- Molecular weight: 651.71
- Purity: 99.44%
- Soluble in DMSO at 25 mg/mL
- Powder storage: -80°C for 2 years, -20°C for 1 year
- In solvent storage: -80°C for 6 months, -20°C for 1 month
- Target: Ser/Thr Kinase
- Related to peptides and enzyme substrates
- Applicable in cancer and cancer metabolism research
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.