Learn More
Medchemexpress LLC Glycine, L-α-aspartyl- | 3790-51-0 | MFCD00037287 | >95.0% | 190.15 g/mol | C6H10N2O5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW129833100MG
Encompass_Preferred
H-Asp-Gly-OH is a cleavable dipeptide linker (Asp-Gly) used in antibody-drug conjugate (ADC) development and conjugation workflows. Supplied as a powder, it is water soluble and has recommended cold storage conditions to maintain stability and activity.
  • Cleavable linker for enzymatic or chemical release in ADCs
  • Short dipeptide sequence facilitating predictable cleavage
  • Powder form suitable for long-term frozen storage
  • Water soluble for straightforward formulation and conjugation
  • Well-characterized molecular properties for reproducible results
Catalog No. 50-002-83259