Learn More
Medchemexpress LLC Glycine, L-y-glutamyl-S-[2-[[4-(dihydroxyarsino)phenyl]amino]-2-oxoethyl]-L-cysteinyl- (TFA) | 00-00-0 | 99.9% | 662.42 g/mol | C20H26AsF3N4O11S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY130588A5MG
Encompass_Preferred
Glutathione arsenoxide TFA is an arsenic-containing derivative of glutathione provided for research use. It acts as a tumour metabolism inhibitor that targets the mitochondrial adenine nucleotide translocator (ANT) and is used to study mitochondrial function and to identify certain cell-surface proteins.
  • Potential anticancer research reagent targeting mitochondrial ANT.
  • Useful for identifying cell-surface proteins such as protein disulfide isomerase.
  • Supplied as a white to off-white solid in trifluoroacetic acid salt form.
  • High purity (≈99.9%) suitable for analytical and biological studies.
  • Documentation available: datasheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-62221