Learn More
Medchemexpress LLC Glycine, N-[(1,1-dimethylethoxy)carbonyl]glycyl-, 2,5-dioxo-1-pyrrolidinyl ester | 29248-48-4 | MFCD00190796 | 97.0% | 329.31 | C13H19N3O7 | 5g
SDP
Supplier:  Medchemexpress LLC HYW8006595G
Encompass_Preferred
Boc-Gly-Gly-NHS ester can be used to selectively attach small molecules to specific amino acid residues on proteins creating conjugates that can be used for a variety of applications in drug discovery and diagnostic assays
Catalog No. 50-002-70354