Learn More
Medchemexpress LLC Glycine, N-[(1,1-dimethylethoxy)carbonyl]-L-phenylalanyl | 25616-33-5 | MFCD00134859 | ≥97.0% | 322.36 g/mol | C16H22N2O5 | 1 G

Supplier: Medchemexpress LLC HYW0488291G
Boc-Phe-Gly-OH is a Boc-protected phenylalanyl glycine derivative used as a protected dipeptide building block for peptide synthesis and related small-molecule synthesis applications.
- Boc-protected phenylalanyl glycine derivative
- Purity ≥97.0%
- Molecular weight 322.36 g/mol
- Chemical formula C16H22N2O5
- Available in 1 G, 5 G, 10 G, 25 G, and 100 G sizes
- Powder storage: -20°C for 3 years, 4°C for 2 years
- Suitable for peptide synthesis and small-molecule preparation
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.