Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycine, N-[(1,1-dimethylethoxy)carbonyl]-L-valylglycine | 45233-75-8 | MFCD00134861 | 97.0% | 274.31 | C12H22N2O5 | 5 G
SDP

Supplier:  Medchemexpress LLC HYW0396955G

Encompass_Preferred

Tert-butoxycarbonyl-L-valylglycine is a Boc-protected dipeptide derivative of glycine supplied as a white to off-white solid for research use. It is used as a building block in peptide synthesis and related biochemical studies where protected amino acid derivatives are required. The material has a molecular weight of 274.31 g/mol and a reported purity of 97.0%.

  • Boc-protected dipeptide suitable for peptide synthesis.
  • White to off-white solid for easy handling.
  • Molecular weight 274.31 g/mol.
  • Molecular formula C12H22N2O5.
  • Purity 97.0% as stated by supplier.
  • Available in small-scale quantities for research use.

Catalog No. 50-002-99761


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.