Learn More
Medchemexpress LLC Glycine, N-(1-oxopropyl)- | 21709-90-0 | MFCD09049674 | 98.0% | 131.13 g/mol | C5H9NO3 | 1 ML

Supplier: Medchemexpress LLC HYW58780310MM/1ML
Propionylglycine is a research-grade N-propionylated glycine supplied as a ready-to-use 10 mM solution in DMSO. It has a reported purity of 98.0%, is provided in a 1 mL vial, and is accompanied by a datasheet and certificate of analysis with solubility and storage guidance for laboratory use.
- Ready-to-use 10 mM solution in DMSO.
- High purity (98.0%) suitable for analytical applications.
- Supplied in a 1 mL vial for small-scale experiments.
- Datasheet and certificate of analysis available.
- Soluble in DMSO; ultrasonic assistance may improve solubility.
- Store under nitrogen: -80°C up to 6 months or -20°C up to 1 month when in solvent.
- Includes CAS number 21709-90-0 and molecular weight 131.13 g/mol for identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.