Learn More
Medchemexpress LLC Glycine, N-[2-[(acetylthio)methyl]-1-oxo-3-phenylpropyl]-, phenylmethyl ester | 81110-73-8 | 99.94% | 385.48 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1739910MM/1ML
Encompass_Preferred
Racecadotril (Acetorphan) is a neutral endopeptidase (NEP) inhibitor and an antidiarrheal agent. Its active metabolite, Thiorphan, inhibits purified NEP activity. Racecadotril is rapidly metabolized in mice and shows efficacy in a rat model of castor oil-induced diarrhea.
Catalog No. 50-003-84598