Learn More
Medchemexpress LLC Glycine, N-(3-pyridinylcarbonyl)- | 583-08-4 | 99.9% | 180.16 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1133531G
Encompass_Preferred
Nicotinuric acid is an acyl glycine and a metabolite of nicotinic acid, appearing as an off-white to light yellow solid. It is classified under ketones, aldehydes, and acids, and originates as an endogenous metabolite.
  • Solid appearance.
  • Off-white to light yellow color.
  • Classified as ketones, aldehydes, and acids.
  • Originates as an endogenous metabolite.
  • Target: Human endogenous metabolite.
  • Pathway: Metabolic enzyme/protease.
  • For research use only.
Catalog No. 50-004-36549