Learn More
Medchemexpress LLC Glycine, N-[(9H-fluoren-9-ylmethoxy)carbonyl]glycyl- | 35665-38-4 | MFCD00190880 | 100.0% | 354.36 g/mol | C19H18N2O5 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0231211G
Encompass_Preferred
Fmoc-Gly-Gly-OH is an Fmoc-protected diglycine reagent with a free C-terminal carboxylic acid. It is used as a cleavable linker building block in antibody-drug conjugate (ADC) synthesis and in peptide synthesis; it couples under standard peptide-coupling conditions and is deprotected under standard Fmoc-deprotection conditions.
  • Fmoc-protected N-terminus with free carboxylic acid for coupling.
  • Cleavable diglycine linker suitable for ADC and peptide synthesis.
  • High reported purity enabling analytical and synthetic applications.
  • Available in laboratory-scale pack sizes, including 1 g.
  • Characterized by CAS 35665-38-4, molecular formula C19H18N2O5, and molecular weight 354.36 g/mol.
Catalog No. 50-002-03115