Learn More
Medchemexpress LLC Glycine, N-[(9H-fluoren-9-ylmethoxy)carbonyl]glycyl | 35665-38-4 | MFCD00190880 | 100.0% | 354.36 g/mol | C19H18N2O5 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW023121100G
Encompass_Preferred
Fmoc-Gly-Gly-OH is an Fmoc-protected diglycine used as a cleavable linker in antibody-drug conjugate synthesis and as a building block for peptide synthesis. It is supplied in multiple pack sizes and is intended for research use; refer to the certificate of analysis for batch-specific purity and testing data.
  • Cleavable linker for antibody-drug conjugate synthesis.
  • Fmoc-protected diglycine suitable for peptide synthesis.
  • High purity as verified by HPLC for each batch.
  • Available in multiple pack sizes, including 100 G.
  • Simple handling and deprotection under standard Fmoc conditions.
Catalog No. 50-002-03142