Learn More
Medchemexpress LLC Glycine, N,N'-[1-(4,5-dimethoxy-2-nitrophenyl)-1,2-ethanediyl]bis[N-(carboxymethyl)- | 117367-86-9 | 98.4% | 25 MG

Supplier: Medchemexpress LLC 25MGHY126789
DM-Nitrophen is a Ca2+ cage that releases Ca2+ when cleaved upon illumination with near-ultraviolet light. It is intended for research use only.
- Releases Ca2+ upon illumination with near-ultraviolet light
- Molecular weight: 473.39
- Formula: C18H23N3O12
- Appearance: Solid
- Color: Light yellow to yellow
- Purity (HPLC): 98.40%
- Storage: Powder -20°C for 3 years; in solvent -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.