Learn More
Hach Company Glycine p-Toluenesulfonic Acid, 25 g
Supplier:  Hach Company 2278024
Encompass
(Glycine PTSA) Protein-TKN standard: 5.665% N, 35.40% crude protein. For checking accuracy and digestion recovery in Total Nitrogen and TKN determinations. 25 g bottle. See 2277800 for the complete set of three standards with increasing resistance to digestion.
Catalog No. 50-199-5714