Learn More
GENOVIS INC GlyCLICK Alexa Fluor 488 | 2mg
Supplier:  GENOVIS INC L1F01200
Encompass
GlyCLICK is a site-specific conjugation technology for IgG using Fc glycan remodeling and click-chemistry. The technology generates stable and homogenous antibody conjugates for IgG from several species and subclasses. Fc-glycan remodeling by complete deglycosylation of the antibody allows for site-specific conjugation using robust click-chemistry, resulting in a degree of label (DOL) or antibody-drug ratio (DAR) of 2.0.
Catalog No. NC1773506