Learn More
Cayman Chemical Glycochenodeoxycholc ACDd4 5mg
SDP
Supplier:  Cayman Chemical 218905 MG
Encompass_Preferred
An internal standard for the quantification of glycochenodeoxycholic acid by GC- or LC-MS
Catalog No. 50-298-7996