Learn More
Cayman Chemical GlycocyamIn 250g
SDP
Supplier:  Cayman Chemical 35792250 G
Encompass_Preferred
A metabolite of glycine and precursor in the biosynthesis of creatine; formed from glycine by GATM and methylated by GAMT to form creatine; levels are altered in individuals with CCDSs
Catalog No. 50-291-0276