Learn More
Medchemexpress LLC Glycocyamine-d2 (guanidinoacetic acid-d2) | 1173020-63-7 | 99.9% | 119.12 | C3H5D2N3O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW021448S5MG
Encompass_Preferred
Glycocyamine-d2 is the deuterium-labeled form of glycocyamine (guanidinoacetic acid), provided as a stable-isotope reference standard for analytical and metabolic research. It is intended for use as an internal standard or tracer in LC-MS, GC-MS, and NMR applications and is supplied in milligram quantities for laboratory use. For research use only.
  • Deuterium-labeled (d2) analog for unambiguous mass spectrometric identification.
  • Suitable as an internal standard for quantitative LC-MS and GC-MS assays.
  • Compatible with NMR and metabolic tracing experiments.
  • High purity (99.9%) for accurate analytical results.
  • Available in small milligram package sizes suitable for analytical workflows.
Catalog No. 50-003-01427