Learn More
Medchemexpress LLC Glycodeoxycholic acid-d4 | 1069132-37-1 | MFCD06658888 | 99.9% | 453.65 | C26H39D4NO5 | 1MG
SDP
Supplier:  Medchemexpress LLC HY125731S1MG
Encompass_Preferred
Glycodeoxycholic acid-d4 is the deuterium-labeled form of the endogenous bile acid glycodeoxycholic acid, provided as a stable isotope internal standard for quantitative analytical methods such as LC-MS and GC-MS. It is intended for use in metabolomics and bioanalytical assays where isotopically labeled standards are required; storage and solubility instructions are provided to preserve analytical integrity.
Catalog No. 50-002-36609