Learn More
Cayman Chemical GlycodeoxycholIc AcId-d4 1mg
SDP
Supplier:  Cayman Chemical 313101 MG
Encompass_Preferred
An internal standard for the quantification of glycodeoxycholic acid by GC- or LC-MS
Catalog No. 50-299-0594