Learn More
Medchemexpress LLC Glycodeoxycholic acid-d5 | 360-65-6 | MFCD00067133 | 454.65 g/mol | C26H38D5NO5 | 1 MG

Supplier: Medchemexpress LLC HYW6539991MG
Deuterium-labeled glycodeoxycholic acid (d5) supplied as an isotope-labeled analytical standard for research and mass spectrometry applications. It provides a labeled analogue of glycodeoxycholic acid to support quantitative assays and metabolic studies.
- Deuterium-labeled internal standard for quantitative analysis.
- Molecular formula: C26H38D5NO5.
- Molecular weight: 454.65 g/mol.
- Offered in small analytical quantities suitable for standards (1 mg).
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.