Learn More
Medchemexpress LLC Glycogen, Mussel | 9005-79-2 | 99.3% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY113511100MG
Encompass_Preferred
Glycogen, Mussel is a glycolytic intermediate and high-energy phosphate that can serve as a form of energy storage in humans, animals, fungi, and bacteria. It is a branched polymer of glucose stored mainly in the liver and skeletal muscle in the human body, supplying glucose to the bloodstream during fasting and to muscle cells during muscle contraction.
  • Functions as an energy storage form
  • Branched polymer of glucose
  • Stored mainly in the liver and skeletal muscle
  • Supplies glucose to the bloodstream during fasting
  • Supplies glucose to muscle cells during contraction
Catalog No. 50-004-36668