Learn More
TARGETMOL CHEMICALS INC glycohyodeoxycholic acid 20MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T815320MG
Encompass_Preferred
Also available in 1 mL and bulk. Please contact Fisher for quotes. glycohyodeoxycholic acid is a major metabolite of the secondary bile acid hyodeoxycholic acid in humans. Purity 99.82%
Catalog No. 50-287-9737