Learn More
Medchemexpress LLC HY-135969 100mg Medchemexpress, Glycol chitosan CAS:123938-86-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY135969100MG
Encompass_Preferred
Medchemexpress, HY-135969 100mg Glycol chitosan CAS:123938-86-3 Glycol chitosan is a chitosan derivative with hydrophilic ethylene glycol branches. Glycol chitosan enhances membrane permeability and leadkage in Glycine max Harosoy 63W cells. Glycol chitosan is water-soluble, biocompatible and biodegradable. Glycol chitosan inhibits E. coli, S. aureus and S. enteritidis growths with MIC values of 4 μg/mL, 32 μg/mL and <0.5 μg/mL, respectively[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1415