Learn More
Medchemexpress LLC Glycolaldehyde-1-13C | 71122-42-4 | 99.0% | 61.04 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY146995S1MG
Encompass_Preferred
Glycolaldehyde-1-13C is a 13C labeled version of Glycolaldehyde. It is intended for research use only and is not sold to patients. Stable heavy isotopes like 13C are incorporated into drug molecules primarily as tracers for quantitative analysis during drug development, with deuteration also being noted for its potential impact on pharmacokinetic and metabolic profiles of drugs.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-62117