Learn More
Cambridge Isotope Laboratories DEUTERIUM (D, 99.8%) (D2,99.6%+HD,0.4%)(IN CODE H-3 CYLINDER), 100 L, 7782-39-0
Supplier:  Cambridge Isotope Laboratories DLM408100
Encompass
DEUTERIUM (D, 99.8%) (D2,99.6%+HD,0.4%)(IN CODE H-3 CYLINDER), 100 L, 7782-39-0. *This item is shipped in a Code H-3 Carbon Steel Cylinder. The fee for the cylinder will need added to the order as a separate charge. Please include part# NC1727200 to the order.
Catalog No. 50-163-1076