Learn More
Medchemexpress LLC Glycolithocholic aci 100mg | 474-74-8 | 433.62 g/mol | C26H43NO4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY116374R100MG
Encompass_Preferred
Glycolithocholic acid is the glycine-conjugated form of lithocholic acid supplied as a high-purity analytical standard for biochemical and analytical applications. It is intended for use as a reference material in LC-MS method development, metabolite identification, and quantitative assays.
  • High purity suitable for analytical use
  • Formulated as a solid standard for easy weighing and solubilization
  • Well suited for LC/MS, method development, and metabolite identification
  • Molecular formula C26H43NO4; molecular weight ~433.62 g/mol
  • Available in small pack sizes for laboratory workflows
Catalog No. 50-002-29473