Learn More
Medchemexpress LLC Glycolithocholic aci 10mg | 474-74-8 | 433.62 g·mol⁻¹ | C26H43NO4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY116374R10MG
Encompass_Preferred
Glycolithocholic acid is a glycine-conjugated secondary bile acid provided as an analytical standard for biochemical and metabolic research. It is used in analytical assays and diagnostic investigations related to gastrointestinal and hepatic conditions.
  • Analytical standard grade suitable for research and assay development.
  • High purity appropriate for chromatography and mass spectrometry.
  • Available in multiple pack sizes and solution formats for experimental flexibility.
  • Soluble in common organic solvents such as DMSO; solution options available.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-003-08085