Learn More
Medchemexpress LLC Glycolithocholic aci 1mg | 2044276-16-4 | 437.65 | C26H39D4NO4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY116374S1MG
Encompass_Preferred
Glycolithocholic acid-d4 is a deuterium-labeled analogue of glycolithocholic acid supplied as an isotope-labeled analytical standard. It is intended for use as an internal standard or tracer in mass spectrometry and metabolic studies, providing consistent mass shifts for quantitative assays.
  • High purity for analytical applications (98.38%).
  • Deuterium-labeled for mass spectrometry internal standard use.
  • Available in small quantities suitable for trace-level assays (500 μg-10 mg).
  • Molecular weight and formula suitable for accurate mass calibration (437.65; C26H39D4NO4).
  • Compatible with common solvents used in LC-MS workflows.
Catalog No. 50-002-29224