Learn More
Medchemexpress LLC Glycolithocholic aci 500ug | 2044276-16-4 | 437.65 | C26H39D4NO4 | 500 UG
SDP
Supplier:  Medchemexpress LLC HY116374S500UG
Encompass_Preferred
Glycolithocholic acid-d4 is the deuterium-labeled isotopologue of glycolithocholic acid supplied as a research-grade reference standard for use as an internal standard in LC-MS/GC-MS and metabolomics studies involving bile acid profiling and biomarker research. It supports accurate quantitation in diagnostic and analytical workflows.
  • Deuterium-labeled internal standard for LC-MS/GC-MS.
  • Reported purity 98.38% suitable for analytical use.
  • Molecular weight 437.65 and formula C26H39D4NO4.
  • Provided in small-quantity research packs (for example, 500 UG) for trace-level analyses.
  • Intended for metabolomics and bile acid biomarker studies.
Catalog No. 50-002-28408