Learn More
Medchemexpress LLC Glycolithocholic aci 5mg | 474-74-8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY116374R5MG
Encompass_Preferred
Glycolithocholic acid is a glycine-conjugated secondary bile acid used as an analytical standard and research reagent for studies of bile acid metabolism and liver and gastrointestinal disease biomarkers. It is supplied as a powdered solid in small analytical quantities suitable for method development and LC-MS reference standards.
  • Used as an analytical standard for bile acid profiling.
  • Glycine-conjugated secondary bile acid and endogenous metabolite.
  • Supplied as a powder; soluble in common organic solvents for analysis.
  • Available in small pack sizes (e.g., 5 mg) appropriate for analytical workflows.
  • Storage recommendations: keep cold, long-term storage at -20 °C for the solid.
Catalog No. 50-002-27157