Learn More
Medchemexpress LLC Glycolithocholic acid 3-sulfate-d4 disodium | 64936-82-9 | 99.6% | C26H37D4NNa2O7S | 1 MG
SDP
Supplier:  Medchemexpress LLC HY141931S11MG
Encompass_Preferred
Glycolithocholic acid 3-sulfate-d4 disodium is the deuterium labeled Glycolithocholic acid 3-sulfate.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Isotope-labeled compound.
  • Deuterium (2H, D).
Catalog No. 50-003-53607