Learn More
Medchemexpress LLC Glycolithocholic acid 3-sulfate disodium | 64936-82-9 | 99.7% | 557.65 | C26H41NNa2O7S | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW5874301MG
Encompass_Preferred
Glycolithocholic acid 3-sulfate disodium is a sulfated, glycine-conjugated secondary bile acid supplied as a research reagent. It functions as a GPR39 agonist, stimulates intracellular calcium signaling, and has reported in vitro inhibition of HIV-1 replication. The compound is provided as a dry sodium salt with defined solubility profiles for preparing stock solutions; handle and store under recommended conditions.
Catalog No. 50-002-84596