Learn More
Cayman Chemical GlycolIthocholIc AcId-d4 1mg
SDP
Supplier:  Cayman Chemical 313081 MG
Encompass_Preferred
An internal standard for the quantification of glycolithocholic acid by GC- or LC-MS
Catalog No. 50-299-0676