Learn More
Cayman Chemical Glycolthocholc ACD 3sulfa 5mg
SDP
Supplier:  Cayman Chemical 352985 MG
Encompass_Preferred
A metabolite of glycolithocholic acid; induces cholestasis and increases biliary cholesterol secretion in rats at 240 µmol/kg; serum levels are increased in patients with cirrhosis, hepatitis, or cholestasis
Catalog No. 50-290-9548