Learn More
Agilent Technologies GLYCOLYSIS STRESS TEST KIT
Supplier:  Agilent Technologies 103020100
Encompass
The Seahorse XF Glycolysis Stress Test Kit reveals key data not evident in endpoint measurements alone, such as glucose uptake and lactate production. Single use reagents for longer shelf life, reagents are suspended in media not DMSO, and there is no need to aliquot, no need to adjust pH.
Catalog No. NC0827640