Learn More
Agilent Technologies AdvanceBio InstantAB GlykoPrep verification standard
Supplier:  Agilent Technologies GKIB965
Encompass
Consists of a library of Human IgG N-linked glycans whose reducing termini are derivatized with the fluorescent dye, InstantAB. The labeled glycans were released using GlykoPrep from the same lot of material that is packaged in the Human IgG GlykoPrep Bioprotein control vials (WS0162) included in the packaging. The library is intended for use as a verification standard for the GlykoPrep platform.
Catalog No. NC1576314