Learn More
QA Bio Inc QA-Bio, Inc. • LudgerTag 2-AB Glycan Labeling Kits
Supplier: QA Bio Inc LTKABA2
2-AB Glycan Labeling Kits • 2-AB (2-aminobenzamide) is one of the most widely used fluorescent labels for glycosylation analysis. The LudgerTag 2-AB Labeling Kit includes all the reagents required to label up to 30 glycans samples.