Learn More
QA Bio Inc QA-Bio, Inc. • Ludger ORELA O-Linked Glycan Deglycosylation Kit
Supplier:  QA Bio Inc LLORELAA2
Encompass
O-Linked Glycan Deglycosylation Kit • The Ludger Liberate Orela Kit has been developed for O-linked deglycosylation of glycoproteins. Released glycans have free reducing terminii to allow fluorescent tagging by reductive amination.
Catalog No. 50-355-835