Learn More
ACROBiosystems Recombinant Protein;Recombinant Ebolavirus EBOV (subtype Zaire,strain Kikwit-95) Envelope Glycoprotein (GP);HEK293;100UG;ZE5,CMV,HCMV,GB,Envelope glycoprotein B,Glycoprotein GP55,UL55,GP55
Supplier: ACROBiosystems ZE5V5221100UG
Recombinant Protein;Recombinant Ebolavirus EBOV (subtype Zaire,strain Kikwit-95) Envelope Glycoprotein (GP);100UG;Ebolavirus GP (virus), His Tag (ZE5-V5221) is expressed from human 293 cells (HEK293). It contains AA Ile 33 - Asp 637 (Accession # AAQ55048.1).;Ebolavirus EBOV(subtype Zaire,strain Kikwit-95) GP, containing GP1 and GP2, is fused with a polyhistidine tag at the C-terminus, and has a calculated MW of 67 kDa. The predicted N-terminus is Ile 33. The reducing (R) protein migrates as 21-23 kDa (GP2-delta) and 110-120 kDa (GP1) in SDS-PAGE .