Learn More
Cayman Chemical Glycopyrrolate 10mg
SDP
Supplier:  Cayman Chemical 1649810 MG
Encompass_Preferred
A mAChR antagonist (Kis = 0.42, 1.77, 0.52, 0.78, and 1.29 nM for the M1-M5 receptors, respectively); induces relaxation of precontracted isolated human bronchi at 0.01, 0.1, or 1 µM; reduces post-prandial gastric antral motility in dogs at 0.01 mg/kg; inhibits salivation in a rat model of pilocarpine-induced sialorrhea (ED50 = 0.74 UG/kg)
Catalog No. 50-297-1377